Sugar Refractometer, 0 to 80%, Sper Scientific
Supplier: Sper Scientific
300003
66214-980EA 789.8 USD
66214-980
Sugar Refractometer, 0 to 80%, Sper Scientific
Refractometers Handheld Refractometers
This unit covers all of the substances of the Sugar-Brix Refractometer 0–32% and Sugar Refractometer, 28–62%, as well as very high sugar content products such as jam, marmalade, and syrup
Its wide range also makes this refractometer a good choice for the testing of any solution with an unknown sugar content. To operate the device, simply place a drop or two of sample on the prism and read the results.
Ordering information: Refractometer is supplied with a hard carrying case, calibration tool, and 5-year warranty.