SP Bel-Art Vaccu-Pette 96-well Format Pipetting Device, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37876-0001
66190-080CS 701.18 USD
66190-080
SP Bel-Art Vaccu-Pette 96-well Format Pipetting Device, Bel-Art Products, a part of SP
Microplate Dispensers
This pipetting device can aspirate or dispense 96 wells in less than 10 seconds.
  • Transparent polycarbonate construction
  • Available in sterile/disposable or autoclavable/reusable models
  • Ideal for efficient washing, dispensing, or aspirating of wells
It speeds up repetitive washing, dispensing, or aspirating of wells by working simultaneously. It has a vacuum outlet at one end, and a short length of 3.2 mm I.D. (1/8") tubing is supplied for syringe connection.
Sterile/disposable model is molded of clear, disposable plastic that can be used only once and cannot be autoclaved or gas sterilized. Autoclavable/Reusable model is molded of polycarbonate, and must be sterilized by autoclaving at no more than 121 °C for 20 minutes at 15 psi prior to use in order to avoid contamination. It cannot be gas sterilized due to the possibility of residual trapped gases.
Simply connect a disposable plastic syringe and withdraw the plunger to transfer liquid. Excellent for simultaneously changing media or for the addition/removal of reagents from a 96-well plate.
Ordering information: Requires a 30 ml or larger disposable sterile syringe (not included).
Delivery information: Complete with a 7.6 cm (3") long piece of 3.2 mm (1/8") tubing for syringe connection. The autoclavable, reusable polycarbonate pipetting device is supplied in a sealed pack with tubing enclosed.