Imhoff Triple Cone Support, Bennett Wood
Supplier: Bennett Wood Specialties
6783
66190-009EA 435.18 USD
66190-009
Imhoff Triple Cone Support, Bennett Wood
Funnels Sedimentation Cones
Wood stand supports 3 Imhoff cones
Constructed so that cone graduation marks fall below the lower support deck for easy reading.