Plankton Nets, LaMotte
Supplier: LaMotte
1063
66175-253EA 223.05 USD
66175-253
Plankton Nets, LaMotte
Samplers Water Samplers
Plankton net are made of cone-shaped nets with nylon cloth to collect plankton in a removable, clear conical graduated tube attached at the end of the net.
  • Net mouth is braced by a sturdy stainless steel ring and harness
  • Two tubes are provided