SP Bel-Art Measure-Matic™ Dispenser, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F11654-0000
66175-154CS 260.65 USD
66175-154
SP Bel-Art Measure-Matic™ Dispenser, Bel-Art Products, a part of SP
Bottle Top Dispensers
Squeezing forces the desired volume of liquid through the tube and into the cup.
  • Polyethylene storage bottle
  • Polypropylene dispensing cup and delivery tube
  • Hollow stopper serves as a dust cover
When pressure is released, excess fluid returns into the bottle for spill-free pouring.
Polypropylene dispensing cup slides onto a polypropylene delivery tube which is set in a low-density polyethylene 500 mL (16 oz.) storage bottle. A supplied hollow stopper serves as a dust cover.