SP Bel-Art® Adjustable Handle Water Sample Dipper, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37807-0000
66175-065EA 274.27 USD
66175-065
SP Bel-Art® Adjustable Handle Water Sample Dipper, Bel-Art Products, a part of SP
Samplers Sample Dippers
Designed for taking samples of water, sewage, wastes, and effluents from lakes, streams, tanks, or ponds, the water sample dipper provides a long reach for otherwise inaccessible spots.
  • Polypropylene construction
  • Adjustable aluminum handle
  • Suited to sampling aqueous solutions
  • Easy to adjust for optimal comfort
A 1000 mL graduated polypropylene pitcher is mounted onto an aluminum tube handle. The handle has three parts, a 91 cm (3') lower section, a 112 cm (44") telescoping section, and an additional 91 cm (3') upper section. Connections are made withMore Product Information