Drum Opener, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
885295
66174-295EA 80.21 USD
66174-295
Drum Opener, Wheaton
Wrenches
Drum opener features a quarter-turn opening action and is cast from a light-weight, durable, sparkless aluminum alloy.
  • For use with hazardous material