Conductivity/TDS Solutions, LaMotte
Supplier: LA MOTTE CHEMICAL PRODUCTS CO
6417-L
66170-814EA 50.89 USD
66170-814
Conductivity/TDS Solutions, LaMotte
Standards Conductivity Standards
Potassium chloride solutions can be used to standardize conductivity meters
TDS values are based on a 0.7 conversion from conductivity.