Test Kits, Sulfite , LaMotte®
Supplier: LA MOTTE CHEMICAL PRODUCTS CO
7175-01
66170-510EA 120.84 USD
66170-510
Test Kits, Sulfite , LaMotte®
Environmental Test Kits
An iodide-iodate titrant oxidizes sulfite to sulfate under acid conditions, until all of the sulfite is reacted
The titrant then reacts with starch to form a blue color signifying the endpoint.