QAC Test Kit, La Motte
Supplier: LA MOTTE CHEMICAL PRODUCTS CO
3043-DR-01
66170-492EA 85.65 USD
66170-492
QAC Test Kit, La Motte
Environmental Test Kits
This kit tests for QAC using the direct reading titrator method and is best suited to higher QAC concentrations.
A masked bromphenol blue indicator is added to the sample and turns green
Range and sensitivity is 0 to 500 ppm/10 ppm alkyl dimethyl benzyl ammonium chloride.
Sodium tetraphenylboron is added to complex the QAC and the color changes to red.