Chlorine Test Kit, Dual Range
Supplier: LA MOTTE CHEMICAL PRODUCTS CO
3314-01
66170-122EA 233.91 USD
66170-122
Chlorine Test Kit, Dual Range
Environmental Test Kits
These Dual Range Chlorine Test Kit tests for high and low ranges of chlorine using the Octa-Slide 2 Comparator.
Delivery information: The kit includes Chlorine DPD 1R and 3R Tablets, Test Tubes 2.5-5.0 - 10.0 mL, plastic, with caps, Octa-Slide Viewer, Chlorine Octa-Slide 2 Bar, 0.1 - 1.0 ppm and 1.0 - 6.0 ppm.