Zobell Solution, ORP Calibrator
Supplier: XYLEM/YSI
061320
66121-265EA 63 USD
66121-265
Zobell Solution, ORP Calibrator
Standards Redox/ORP Standards
Zobell solution is used for calibrating ORP/Redox on several YSI instruments.
A convenient temperature chart with corresponding ORP values is included for simple calibration value inputs. Concentrated powder makes 125, 250, or 500 ml of calibration standard.