AMCO Clear® Primary Turbidity Standards, GFS Chemicals
Supplier: GFS CHEMICALS INC
85151
85047
85040
85034
85075
85007
85011
85016
85024
85027
85031
85043
85050
85056
85062
85068
85074
85093
85123
85141
85159
86110
85009
85005
85014
85018
85030
85026
85051
85063
85069
85092
85122
85142
85150
85160
85164
85166
66115-038EA
138.57
USD
66115-038
66115-136
66115-138
66115-140
66115-152
66115-000
66115-002
66115-004
66115-006
66115-008
66115-010
66115-014
66115-016
66115-018
66115-020
66115-022
66115-024
66115-026
66115-028
66115-034
66115-042
66115-046
66115-128
66115-130
66115-132
66115-134
66115-142
66115-144
66115-146
66115-148
66115-150
66115-154
66115-156
66115-162
66115-166
66115-170
66115-172
66115-174
AMCO Clear® Primary Turbidity Standards, GFS Chemicals
Standards
Turbidity Standards
EPA-approved primary turbidity standards for calibrating turbidimeters and nephelometers
One year shelf life.
Ready to use at their stated value without mixing or diluting. Composed of submicron solid polymer beads suspended in ultra-pure water. Guaranteed accuracy within 1%. Safe, non-toxic, non-carcinogenic alternative to Formazin. Available for non-ratio and ratio meters.
Certifications: Approved by EPA, Standard Methods, ASTM, ASBC, and EBC. Traceable to NIST 1690, 1691.