Target® Syringe Filters, PVDF, Thermo Scientific
Supplier: Thermo Scientific
F2500-6 F2500-5 F2513-5 F2504-5 F2513-6 F2504-6
66064-397PK 485.15 USD
66064-397 66030-847 66030-867 66030-880 66064-433 66064-832
Target® Syringe Filters, PVDF, Thermo Scientific
Filters Syringe Filters
PVDF (polyvinylidene difluoride) is a hydrophilic, solvent-resistant membrane with low levels of UV absorbing extractables.
  • Low nonspecific binding
  • Use for HPLC sample filtration and general biological filtration
  • PVDF is also a low protein binding membrane
  • Enhanced female Luer-lock inlet; male Luer slip outlet
Housing manufactured from medical grade, virgin polypropylene. The 4 mm filters can be used for volumes less than 2 ml. The 0.2 µm pore size filter can be used for 'cold' sterilization.