National™ Vial Racks for 4 ml Vials, Thermo Scientific
Supplier: Thermo Scientific
C4015-25
66030-546CS 521.2 USD
66030-546
National™ Vial Racks for 4 ml Vials, Thermo Scientific
Racks Chromatography Vial Racks
Store vials efficiently with National 4 ml screw top vial racks, which are stackable for even more space savings.
The polypropylene vial racks feature alphanumeric indexing for easier vial identification and are resistant to most solvents.
Order Now
SPECIFICATIONS
Environmentally Preferable