Target DP™ Glass Screw-Thread Vials, 9-42, Thermo Scientific
Supplier: Thermo Scientific
C4000-LV2W C4000-LV1 C4000-1 C5000-680
66030-996PK 309.99 USD
66030-996 66030-998 66031-045 66064-858 66064-910 66064-912 66064-914 66064-952 89187-282 97054-232 97054-234 97054-240 97054-242 66030-000 66030-006 66030-990 66030-992 66030-994 89235-910 89235-912 89235-918 82428-552 10766-770 10781-888
Target DP™ Glass Screw-Thread Vials, 9-42, Thermo Scientific
Vials
Nearly eliminate septa push-through and improve sealing with Advanced Vial Closure System technology. This innovation not only prevents septa push-through, but provides a stop point for the cap to prevent over - or under-tightening and provide an optimal, consistent seal every time. Vials are made from superior quality 33 expansion borosilicate clear Type 1, Class A glass, and meet all US, EU, and JPN Pharmacopia standards.
  • Closure system prevents septa push-through
  • Wide neck opening
Fill vials easily with the wide neck opening; requires 6 mm diameter micro-inserts.
Thermo Scientific National Preassembled Target DP Kits include 100 2 ml vials with pre-attached caps and septa. Packaged in convenient vial trays with clear covers or in polybags.
Get maximum sample extraction without the need for separate inserts by using Microsampling and High Recovery Vials. Identify sample and volume with graduated, write-on patch provided on certain vials. Obtain maximum recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional). The polyspring microseal inserts self-align and provide a cushion against needle contact. Precision point inserts minimize residual sample loss. Pulled point inserts provide an economical choice for noncritical applications. Vials provide the sealing and performance of a crimp top vial. The Sure Stop vial design incorporates a definite stopping point in the vial finish and removes all subjectivity in achieving optimal seal compression closing a vial. Prevents over- or under-tightening of closures and gives an optimal seal every time. Recommended for most brands of autosampler.