SP Bel-Art Scintillation Vial Rack, Polypropylene, 24-Position, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18512-0000
66022-503CS 264.79 USD
66022-503
SP Bel-Art Scintillation Vial Rack, Polypropylene, 24-Position, Bel-Art Products, a part of SP
Racks Scintillation Vial Racks
Rugged, stable polypropylene rack holds 24 scintillation vials (30 mm) for storage or preparation of cocktails.
  • Rugged and stable
  • Polypropylene construction
  • Autoclavable
Scintillation vial rack is resistant to corrosion and convenient for storage, handling, and sample preparation.
Shelf height: 4.4 cm (13/4")