VWR® Solvent-Saver™ Scintillation Vials, Borosilicate Glass, with Screw Cap
Supplier: VWR International
66022-300
66022-300CS 716.46 USD
66022-300
VWR® Solvent-Saver™ Scintillation Vials, Borosilicate Glass, with Screw Cap
Vials
These Solvent-Saver vials feature small vial dimensions to permit substantial reduction in the quantity of solvent required with resultant savings in solvent costs.
  • Smaller overall dimensions
  • Reduced amount of solvent required
The white urea screw cap has a cork-backed foil liner and the top of the cap is suitable for marking.
Ordering information: Replacement caps available (sold separately).
Packaging: Vials are clean-packed in trays of 200, with 5 trays per case. Caps are clean-packed in plastic bags of 200 pieces.