Dissolution Cannula, SUN-SRi
Supplier: Thermo Scientific
401 350
66016-114EA 212.36 USD
66016-114
Dissolution Cannula, SUN-SRi
Cannulae
Cannula is manufactured of Type 316 stainless steel and is angled to allow easy access to samples
A Luer-lock connection provides easy attachment to syringes and a tip sleeve allows secure filter attachment. Sheath included. For 900mL volume.