VWR® Shell Vial and Plug Cap
Supplier: VWR International
66015-702A
66015-702ACS 176.81 USD
66015-702A
VWR® Shell Vial and Plug Cap
Vials
Designed for use in Alcott®, Waters®, Gilson®, or Shimadzu® autosamplers, the clear snap-plug closures of these shell vials can be pierced with standard autosampler syringes without vacuum formation.
  • Polyethylene plug with starburst center design eases syringe needle penetration
  • Convenient vial kits include equal quantities of vials and caps
  • Provides an economical option to autosampler vials
Manufactured from borosilicate glass. Vials and snap-plug closures packed separately in clear-pack, three chamber trays.