Black Phenolic Screw Cap, Open-Top, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
240680
66012-658CS 300.59 USD
66012-658
Black Phenolic Screw Cap, Open-Top, Wheaton
Closures Bottle Closures
Threaded cap is flanged with an open top and a butyl septum.
These black phenolic screw cap, open-top are made of phenolic resin.