WHEATON® Preassembled Open Top Caps with Septa, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
242245
66012-343CS 160.99 USD
66012-343
WHEATON® Preassembled Open Top Caps with Septa, DWK Life Sciences
Closures Chromatography Vial Closures
Natural PP cap with 10 mil septa. For use with standard opening vials (12×32 mm).