Septa for 8-425 Screw-Thread Vial Caps, Thermo Scientific
Supplier: Thermo Scientific
C4013-61 C4013-60 C4013-32
66011-192PK 81.3 USD
66011-192 66064-280 66065-154 69400-090 66011-188 66030-970 72720-109 72720-108
Septa for 8-425 Screw-Thread Vial Caps, Thermo Scientific
Septa
Septa for 8-425 screw thread caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of HPLC, GC and IC applications.
  • PTFE Disk septa are chemically inert
  • PFTE red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin gauge needles