Gray Butyl Rubber Stoppers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
73828A-21 73828-13
66004-008CS 486.99 USD
66004-008 66010-865
Gray Butyl Rubber Stoppers, Kimble Chase
Stoppers Rubber Stoppers
Lyophilization style, slotted stopper for aluminum seals.
  • Two-leg style reduces possibility of legs sticking together
Gray high grade butyl rubber.