KIMAX® Sample Vials, Borosilicate Glass, with PTFE-Lined Screw Cap, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
60940A-8 60940A-12 60940A-16 60940A-24 60940A-2 60940A-4
66009-984CS 320.84 USD
66009-984 66009-985 66009-986 66009-987 66009-988 66009-981
KIMAX® Sample Vials, Borosilicate Glass, with PTFE-Lined Screw Cap, Kimble Chase, DWK Life Sciences
Vials
With PTFE-lined solid-top screw caps attached to vials.
The 16 ml size complies with EPA 600 Series Methods. Made of KG-33 borosilicate glass.