Ampule Snapper, Disposable, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
177105
66009-125CS 43.51 USD
66009-125
Ampule Snapper, Disposable, WHEATON®, DWK Life Sciences
Ampule Openers
Disposable ampule snapper protects the user against glass splinters and specially designed for 1 to 2 ml samples.
It prevents cross contamination between samples.