Nalgene® Sterile Specimen Cryogenic Vial with Screw Closure, Thermo Scientific
Supplier: Thermo Fisher Scientific
5005-0015
66008-740CS 226.36 USD
66008-740
Nalgene® Sterile Specimen Cryogenic Vial with Screw Closure, Thermo Scientific
Vials
Nalgene® Sterile Specimen Cryogenic Vials are designed for storage of solid specimens.
They feature a linerless closure, as well as a wide-mouth, shoulderless vial that allows easy access to sample with forceps. The vials are for use in mechanical freezers only.
Certifications: This product is in compliance to Directive 98/79/EC of the European Parliament and of the Council of 27 October 1998 on in vitro diagnostic medical devices. Certified non-cytotoxic and non-pyrogenic.
Packaging: Packaged 5 vials per easy open bag.