Nalgene® CryoBox™, SYSTEM 100™, Thermo Scientific
Supplier: Thermo Fisher Scientific
5026-1010
66008-714CS 188.62 USD
66008-714
Nalgene® CryoBox™, SYSTEM 100™, Thermo Scientific
Boxes Cryoboxes
These CryoBox™ containers are designed for ultralow temperature storage of one hundred SYSTEM 100™ cryogenic vials (see 66008-708 series), other vials of similar size, externally-threaded vials, and most internally-threaded vials
They increase freezer storage capacity by 23% over boxes that hold 81 vials. Constructed of polycarbonate. 10 x 10 array. A transparent lid has printed grid for inventory purposes. It permits viewing of contents and is keyed to prevent misalignment.
Temperature Range: –196 to 121 °C (–321 to 250 °F)