SP Bel-Art Beta Splash Shield, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F24975-0000
66001-106EA 1051.27 USD
66001-106
SP Bel-Art Beta Splash Shield, Bel-Art Products, a part of SP
Shields Radiation Shields
The Beta Splash Shield is made from 9.5 mm (3⁄8") clear acrylic for protection from Beta emitters and splashes from lab procedures.
  • Clear acrylic construction
  • Excellent visibility of work surface
  • Nonskid feet
  • Can be used while seated or standing
The shield provides a large viewing area and flat work surface.
The unique lower bend and wider top of this shield allow greater visibility of the work surface. May be used from either a standing or a sitting position. The gentle bend at 21.6 cm (81/2") from the base allows the technician to observe the working surface from a comfortable sitting position. Nonskid feet. Fabricated from 9.5 mm (3/8") clear acrylic.