Nalgene® Check Valve, High-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
6120-0010
65551-000CS 1076.7 USD
65551-000
Nalgene® Check Valve, High-Density Polyethylene, Thermo Scientific
Valves Vacuum Valves
For use in vacuum lines and to prevent back pressure in filter flasks
Not for use with liquids. Tubing connections are 3/4" long and fit 1/45/16" I.D. tubing. Withstands temperatures up to 82.2°C (180°F). Works best between 254–508mm Hg (10–20in. Hg).