Anti-SIK2 Mouse Monoclonal Antibody [clone: S15G10]

Supplier: Biolegend
636702
636702-BLEA 257.8 USD
636702-BL
Anti-SIK2 Mouse Monoclonal Antibody [clone: S15G10]
Antibodies
Purified anti-SIK2 [S15G10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IF, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested working dilution is 1:250. For immunofluorescence microscopy, use a dilution range of 1:100 to 1:400. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SIK2
Clonality: Monoclonal
Clone: S15G10
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR