Heidolph® Titramax Vibrating Platform Shaker Package, Heidolph North America
Supplier: Heidolph NA, LLC
036130500
63400-134EA 9004 USD
63400-134
Heidolph® Titramax Vibrating Platform Shaker Package, Heidolph North America
Shakers and Mixers Microplate Shakers
The Vibrating Platform shaker is best used for research which requires ridged or gentle shaking for small samples on microtiter plates or test tubes
The Heating modules has a heating capacity of 300W which allows for quick temperature adjustments. Digital temperature setting up to 65C and separate display for actual temperature.
The Flat hood is for small vessels with a total height of 163mm, This is recommended especially for microtiter plates, culture plates, small flaks, media bottles and Erlenmeyer flasks up to 100ml. Transparent and non-fogging PETG material allows for sample viewing and easy cleaning.
With a capacity of 6 mirotiter plates it increases your throughput by 50%. The 1.5mm vibration orbit perfoms gentle mixing with best results using small sample volumes
Ordering information: This package includes one Titrmax 1000 vibrating platform shaker, One heating module and one flat incubator hood