UVP UVX™ Digital Radiometer, Analytik Jena
Supplier: Analytik Jena US
97-0015-02
63220-114EA 1512.63 USD
63220-114
UVP UVX™ Digital Radiometer, Analytik Jena
Light Meters
Hand-held radiometer is a state-of-the-art measuring device ideal for obtaining accurate and repeatable UV irradiance measurements. Built to withstand day-to-day use in laboratory and production environments.
Compact, portable, easy-to-operate unit features automatic self-test circuitry, read/hold switch, low battery indicator, and a 9.1 cm (319/32") window with LCD readout
Port enables unit to be connected to a chart recorder for continuous monitoring. Unit operates for 120 hours on a 9 V battery (not included).
Unit accommodates three interchangeable sensor probes (sold separately) sensitive in the 254 nm, 365 nm, and 302 nm regions of the ultraviolet spectrum. Probes feature 0.9 m (3') electrically shielded cables and are capable of measuring sensitivity ranges from 0 to 20 mW/cm2, 0 to 2000 µW/cm2, and 0 to 200 µW/cm2. A 10:1 attenuator screen accessory (sold separately) extends UV measurement range to 200 µW/cm2.
Certifications: CE marked. A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology (NIST) and Analytik Jena Standards.
Ordering information: Supplied with a hinged hard plastic carrying case. Sensor probes sold separately.