Pliers, Form Dog Leg, Excelta
Supplier: Excelta
500-13B-US 500-88-US 500-89B-US 907-89 907-89B 900-TXC 500-92-US 900-11 500-13B-US 500-89-US
63042-410EA 642.86 USD
63042-410 63042-418 63042-420 63043-042 94024-396 89411-970 75880-612 75880-654 76251-154 76267-620 76267-784
Pliers, Form Dog Leg, Excelta
Pliers
These are designed to form a consistent stand off and cut the lead to length on most popular radial lead components.
  • Forms 'dog leg' on component leads
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
This forming plier/cutter will form a 'Gull Wing' and cut multiple leads to length on various types of QFP and SOIC packages. It features an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. These tools are made of carbon tool steel with flame hardened cutting edges.