Three Star™ Tweezers, Anti-Crush Devices, Excelta Corp®
Supplier: Excelta
52A-SA 53C-SA
63042-518EA 100.13 USD
63042-518 63042-570
Three Star™ Tweezers, Anti-Crush Devices, Excelta Corp®
Forceps
Outfitted with an 'anti-crush' devise, these tweezers define the amount of pressure put on a delicate component.
  • Anti-crush device protects delicate items
  • Cleanroom safe
  • Made of anti-magnetic stainless steel
The tweezer tips collapse when excessive force is placed on the tweezer.
These two tweezers are designed for protecting delicate items. The anti-crush device allows the tweezer tips to collapse when too much pressure is applied. They are made of anti-magnetic Neverust® stainless steel.