Tweezers, Straight Fine Tip, Style 3C, Excelta Corp®
Supplier: Excelta
3C 3CB-S 3C-PI 3C-S 3C-SA-ET 3C-SA-PI 3C-SA-SE 3C-SE 3C-S-PI 3C-S-SE 3C-SA-PI-ET 3C-SA 3C-S 3C-SA-ET 3C-SA-R 3C-SA-MZ
63042-108EA 106.94 USD
63042-108 63042-110 63042-118 63042-120 63042-122 63042-124 63042-128 63042-130 63042-132 63042-136 63042-138 89411-772 76238-684 76238-686 76240-490 76243-424 76471-468 76471-470 76471-674
Tweezers, Straight Fine Tip, Style 3C, Excelta Corp®
Forceps
Style 3C is ideal for precision work, including under magnification.
  • Ideal for working under magnification
  • Available in three different materials
  • Available in three different qualtiy grades
  • Ergonomic handles are standard option
  • Teflon coated tip option for non-marring
The 'Style 3C' series straight very fine precision pointed tweezer is designed for work under magnification. The overall length is 4.25".The different options offered are ergonomic handles or teflon coated tips (to prevent scratching).depending on your application. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).