Two Star™ Hemostats, Excelta Corp®
Supplier: Excelta
36-SE 37-SE 38-SE 35-A-SE 39-SE 40-SE
63042-028EA 14.72 USD
63042-028 63042-030 63042-050 63042-052 63042-070 63042-072 63042-088 63042-090 89411-716 75880-830 75880-832 75880-834 75880-836
Two Star™ Hemostats, Excelta Corp®
Forceps
Ideal for clamping parts for inspection, these hemostats come in various sizes and some include vinyl-coated handles.
  • Stainless steel construction
  • Locking for long-term holding
  • Available with vinyl-coated handles
These stainless steel locking hemostats offer serrated jaws for positive gripping. Each is available with vinyl-coated handles (PH).