Replacement scalpel blades, Excelta Corp®
Supplier: Excelta
177-11
63041-316EA 2.29 USD
63041-316 63041-318 63041-320 63041-322 75880-950
Replacement scalpel blades, Excelta Corp®
Scalpels and Scalpel Assemblies Scalpel Blades
These scalpel blades are all stainless steel (AISI 440) except the 181-22 which is carbon steel.