BD INTRAMEDIC™ Polyethylene Tubing, BD Medical
Supplier: BD
427405 427410 427415 427420 427425 427430 427435 427440 427445 427450 427401 427406 427411 427416 427421 427426 427431 427436
63018-646EA 48.82 USD
63018-646 63018-668 63018-670 63018-704 63018-726 63018-748 63018-770 63018-781 63018-806 63018-828 63019-004 63019-026 63019-048 63019-070 63019-081 63019-106 63019-128 63019-150
BD INTRAMEDIC™ Polyethylene Tubing, BD Medical
Tubing Transfer Tubing
Polyethylene tubing is flexible, but rigid enough for applications using vacuum and pressure.
Polyethylene contains no additives or plasticizers. Nonsterile, nonradiopaque, and non-tissue-reactive. Waxy white, translucent, tasteless, odorless. Durable over a wide temperature range, up to 104 °C (219 °F).
Ordering information: Supplied in 10' and 100' lengths.