Tygon® SPT-3370 IB Food and Beverage Transfer Tubing
Supplier: Saint Gobain Life Sciences
AHJ001718 AHJ001719 AHJ001720 AHJ001721 AHJ001722 AHJ001724
63014-420PK 483.8 USD
63014-420 63014-422 63014-424 63014-426 63014-428 63014-430 63014-432
Tygon® SPT-3370 IB Food and Beverage Transfer Tubing
Tubing Transfer Tubing
Natural translucent silicone tubing is ideal for hot fill lines, food handling, stirring vessels, fermenter lines, laboratory, chemical, and resin feeds, viscous and adhesive materials, and D.I water transfer.
  • Braid reinforcement increases pressure resistance
  • Provides flexibility, durability, and chemical and temperature resistance
  • Durometer hardness: Shore A, 70, tensile strength: 1200 psi, autoclavable
  • Platinum-curing process helps ensure the highest level of biocompatibility available
The ultra-smooth inner surface reduces the potential for particle entrapment and helps maintain fluid integrity throughout sensitive fluid transfer or processing. Withstands repeated CIP and SIP sterilization. Minimum operating temperature: –80 °C (–112 °F). Maximum operating temperature: 160 °C (320 °F).
Certifications: Meets USP Class VI and FDA criteria.
Ordering information: Supplied in 50' coil lengths.