Versilon™ F-5500-A Cleaning Fluid Line and Solvent Recovery Tubing
Supplier: Saint Gobain Life Sciences
AGN00022
63014-305EA 287.34 USD
63014-305 63014-327 63014-360 63014-382 63014-407
Versilon™ F-5500-A Cleaning Fluid Line and Solvent Recovery Tubing
Tubing Transfer Tubing
Black Versilon™ F-5500-A is an opaque tubing for use in severe environments where other tubing fails, such as peristaltic pump tubing for extremely corrosive materials, solvent recovery systems, dry-cleaning fluid lines, and process monitoring equipment.
Resistant to ozone, sunlight, weathering, corrosive chemicals, oils, fuels, solvents, aliphatic, and aromatic hydrocarbons. Withstands temperatures from –51 to +204 °C (–60 to +400 °F). Flexible and resilient. Durometer hardness: Shore A, 60. Tensile strength: 1400psi at 23 °C (73 °F). Not for use with foods, beverages, or drugs.
Ordering information: Available in 50' coil lengths.