Versilon™ C-210-A Multipurpose Fuel and Oil Transfer Tubing
Supplier: Saint Gobain Life Sciences
AEM02053
63011-451EA 208.77 USD
63011-451 63011-455 63011-457 63011-475 63011-478 63011-493 63011-489
Versilon™ C-210-A Multipurpose Fuel and Oil Transfer Tubing
Tubing Transfer Tubing
Rigidly controlled manufacturing processes ensure that Tygothane® Precision Polyurethane Tubing the flexible polyurethane tubing has consistently tight tolerances from lot to lot
Versilon™ C-210-A Multipurpose fuel and oil transfer tubing provides consistently tight dimensional tolerances, excellent abrasion and tear resistance, and excellent resistance to oils, greases, and fuels. It retains flexibility in sub-zero environments.
Made of tough ester-based polyurethane, Versilon™ C-210-A Multipurpose fuel and oil transfer tubing's clarity, high tear strength, and excellent abrasion resistance make it ideal for applications including fuel and lubricant lines, pneumatic lines, abrasive product transfer, and cable jacketing. It also offers exceptional resistance to oils, greases, fuels, and many chemicals. Able to withstand rugged daily use, Versilon™ C-210-A Multipurpose fuel and oil transfer tubing resists weathering and can be safely used in temperatures ranging from –73 to +93 °C (–100 to +200 °F).
While many rubber and plastics materials exhibit resistance to certain solvents, oils, and chemicals, Versilon™ tubing will resist a much wider range of substances. Plasticizer extraction leading to embrittlement is one of the most frequent causes of failure when flexible tubing is exposed to harsh chemicals. Versilon™ Multipurpose fuel and oil transfer tubing is plasticizer-free and remains flexible even when cycled through temperature extremes. The tubing's precision tolerances and high elasticity provide the user with an easy, secure, and worry-free attachment to fittings.
Certifications: Tubing meets FDA 21 CFR, 177.1680 and 177.2600 criteria for food and beverage use. Also available in ether-based and medical grade formulations.
Ordering information: Versilon™ C-210-A Multipurpose fuel and oil transfer tubing is supplied in 100' lengths.