PYREX® Distilling Adapter, Corning
Supplier: Corning
8950-24
62939-996CS 263.3 USD
62939-996
PYREX® Distilling Adapter, Corning
Distillation Components Distillation Adapters
These PYREX® 60° angle distilling connecting adapters have a 24/40 Standard Taper outer joint at the upper end and inner 24/40 Standard Taper joints and on the sidearm and lower joints
This tube also serves as a replacement part for organic chemistry kit No. 6949