SP Bel-Art Cleanware™ Label Remover, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F17077-0000
62866-188EA 66.41 USD
62866-188
SP Bel-Art Cleanware™ Label Remover, Bel-Art Products, a part of SP
Cleaning Agents Detergents and Surface Cleansers
Cleanware™ Label-Off™ Label Remover quickly removes labels, including the gummy residue other cleaners leave behind.
  • Quick and easy removal of labels
  • No residue left behind
The kit contains 59 ml (2oz.) of label adhesive solvent in a pump spray with cap, along with 20 lint-free, 10.2 cm (4") square wipes.
Ordering information: No air shipment.