SP Bel-Art Adjustable Microplate Tilting Stand, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18938-0000
62866-107EA 245.15 USD
62866-107
SP Bel-Art Adjustable Microplate Tilting Stand, Bel-Art Products, a part of SP
Well Plate Stands and Well Orienters
Make precise additions of liquid components to microplate wells by adjusting plates to the most convenient angle.
  • Epoxy coating
  • Clear acrylic platform
  • Durable steel base
  • Fully adjustable for providing convenient microplate angles
  • Compatible with all sizes of microplates
The Tilting Stand helps reduce errors in non-manifold additions of assay components to a multiwell microplate, including expensive, hazardous, or radioactive materials
An epoxy-coated steel base provides stability, and the hand-adjustable clamping screws maintain the desired angle.More Product Information