Anti-TRIM Mouse Monoclonal Antibody [clone: TRIM-4]

Supplier: Biolegend
627202
627202-BLEA 329.9 USD
627202-BL
Anti-TRIM Mouse Monoclonal Antibody [clone: TRIM-4]
Antibodies
Purified anti-TRIM [TRIM-4]; Isotype: Mouse IgG2a; Reactivity: Human, Mouse; Apps: WB, IF, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TRIM
Clonality: Monoclonal
Clone: TRIM-4
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR