Chemware® Tray with Cover, Saint Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
D1069670
62670-052EA 267.9 USD
62670-052
Chemware® Tray with Cover, Saint Gobain Performance Plastics
Trays/Dishes/Pots Laboratory Trays/Dishes
The chemically resistant trays are excellent for corrosive and solvent baths.
  • Compatible with temperatures between –370 to 500°F (–200 to 260°C)
  • Size is ideal for staining standard microscope slides
  • Cover prevents evaporation of liquid contents