Nalgene® Pans, High-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
7120-0010 7120-0020
62662-875CS 278.24 USD
62662-875 62662-886
Nalgene® Pans, High-Density Polyethylene, Thermo Scientific
Trays/Dishes/Pots Laboratory Trays/Dishes
Pans are constructed of high-density polyethylene for rigidity and durability
Use for soaking labware or as an ice bath. With handles for easy carrying. Tapered for nesting.