Nalgene® Instrument/Pipet Sterilizing Pan, Filled Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
6910-0618
62662-751CS 613.14 USD
62662-751
Nalgene® Instrument/Pipet Sterilizing Pan, Filled Polypropylene, Thermo Scientific
Trays/Dishes/Pots Instrument Trays/Dishes
Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research, where disinfecting and subsequent autoclaving of pipets are required before cleaning or disposal
Horizontal design allows pipets to be fully immersed in disinfectant. Plastic construction cuts down on breakage and chipping of glass pipet tips. Cover fits into groove of pan to minimize spillage of hazardous materials. Pan is resistant to aggressive disinfectants. Can also be used to sterilize small labware instruments and utensils. Autoclavable.
Stackable, lightweight, quiet, and less expensive than stainless steel.
Certifications: Meets OSHA Standard 29CFR Part 1910.1030 for use as protection against bloodborne pathogens.