AkroBins®, Akro-Mils
Supplier: Akro-Mils
30239BLUE
62661-530CS 627.02 USD
62661-530
AkroBins®, Akro-Mils
Boxes Storage Boxes
Optimize your storage space.
  • Rust, Water, and Corrosion-Proof
  • Industrial-Grade Polymer Bins
  • Autoclavable up to 250⁰F
  • Wide Range of Labeling Options Available
AkroBins are the industry-leading hanging and stacking plastic storage bins.
Heavy-duty, industrial-grade polymer bins stack together or hang from Akro-Mils louvered racks and panels, rail racks, storage cabinets, and louvered carts for high-density storage. AkroBins are corrosion proof and unaffected by weak acids and alkalis. Dividers, lids, and a wide range of labeling options are available.